(-)-Dibenzoyl-L-tartaric acid, anhydrous 99%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
(2R,3R)-2,3-Bis(benzoyloxy)succinic acid
A16181.14
A16181.22
A16181.36
A16181.14EA
358
DKK
A16181.14
A16181.22
A16181.36
(-)-Dibenzoyl-L-tartaric acid, anhydrous 99%
(-)-Dibenzoyl-L-tartaric acid
Purity: 99%
CAS No.: 2743-38-6
Molecular Formula: C18H14O8
Formula Weight: 358.31
Melting Point: 154-156°
MDL No.: MFCD00003074
Formel:
C₁₈H₁₄O₈ Molekylvægt: 358.31 g/mol Smeltepunkt: 154…156 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00003074 CAS-nummer: 2743-38-6 EINECS: 220-374-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...