2-Chloro-N,N-diethylacetamide ≥97%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

A13691.06 A13691.14 A13691.22
A13691.06EA 376 DKK
A13691.06 A13691.14 A13691.22
2-Chloro-N,N-diethylacetamide ≥97%
2-Chloro-N,N-diethylacetamide

Purity: 97% CAS No.: 2315-36-8 Molecular Formula: C6H12ClNO Formula Weight: 149.62 Flash Point: 113°(235°F) Refractive Index: 1.4700 Boiling Point: 123-124°/15mm Beilstein Reference: 605866 UN No.: 2810 MDL No.: MFCD00000928

Formel: C₆H₁₂ClNO
Kogepunkt: 123…124 °C (15 mmHg)
Massefylde: 1,087
Flammepunkt: 113 °C (235 °F)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00000928
CAS-nummer: 2315-36-8
EINECS: 219-019-2
UN nummer: 2810
ADR: 6.1,II

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR