Cresol red sodium salt
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer: Cresol Red sodium, Sodium 4-(3-(4-hydroxy-3-methylphenyl)-1,1-dioxido-3H-benzo[c][1,2]oxathiol-3-yl)-2-methylphenolate
B21361.14 B21361.22
B21361.14EA 805 DKK
B21361.14 B21361.22
Cresol red sodium salt
Cresol red sodium salt
CAS No.: 62625-29-0 Molecular Formula: C21H17NaO5S Formula Weight: 404.42 Melting Point: ca 250° dec. MDL No.: MFCD00001618
Formel: C₂₁H₁₇NaO₅S
Molekylvægt: 404.42 g/mol
Smeltepunkt: ∼ 250 °C (dec.)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00001618
CAS-nummer: 62625-29-0
EINECS: 263-654-8