Cresol red

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: o-Cresolsulphonphthalein, 3,3-Bis(4-hydroxy-3-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide

A17243.14 A17243.22
A17243.14EA 623 DKK
A17243.14 A17243.22
Cresol red
Cresol red

CAS No.: 1733-12-6 Molecular Formula: C21H18O5S Formula Weight: 382.44 Melting Point: 300° Beilstein Reference: 343399 Merck Reference: 14,2578 MDL No.: MFCD00005878

Formel: C₂₁H₁₈O₅S
Molekylvægt: 382.43 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00005878
CAS-nummer: 1733-12-6
EINECS: 217-064-2

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR