3-Bromo-5-(trifluoromethyl)pyridine 95%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
H25823.06
H25823.03
H25823.06EA
4620
DKK
H25823.06
H25823.03
3-Bromo-5-(trifluoromethyl)pyridine 95%
3-Bromo-5-(trifluoromethyl)pyridine
Purity: 95%
CAS No.: 436799-33-6
Molecular Formula: C6H3BrF3N
Formula Weight: 226.00
Flash Point: 85°(185°F)
Melting Point: 44-46°
Boiling Point: 65-66°/19mm
MDL No.: MFCD04972700
Formel:
C₆H₃BrF₃N Kogepunkt: 65…66 °C (19 mmHg) Smeltepunkt: 44…46 °C Flammepunkt: 85 °C (185 °F) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD04972700 CAS-nummer: 436799-33-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...