Dipentyl phthalate, Supelco®

Leverandør: Merck
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Di-n-pentyl phthalate

M-Clarity™ quality level = MQ100

80154-1ML 80154-5ML
80154-1MLEA 517 DKK
80154-1ML 80154-5ML
Dipentyl phthalate, Supelco®
Dipentyl phthalate
Formel: C₁₈H₂₆O₄
Molekylvægt: 306.41 g/mol
MDL Number: MFCD00041934
CAS-nummer: 131-18-0
EINECS: 205-017-9
UN nummer: 3082
ADR: 9 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b55a2f3c4a5e31baea1baadc9e6827de6212f6d3&vid=4dc14ab2a7463ee9467291051c6c58f4f945e692&prid=web&clid=SAPDAM

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR