Surgical blades, Cygnetic®
Leverandør: SWANN MORTON
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
5301 5303 5305 5306
SWAN5301EA 735 DKK
SWAN5301 SWAN5303 SWAN5305 SWAN5306
Surgical blades, Cygnetic®
Skalpeller Skalpelblade
These stainless steel blades are thicker than a standard blade thus offering additional rigidity and strength when used in conjunction with the unique Cygnetic ® stainless steel handle.
Ideal for Orthopaedic procedures where the surgeon may exert excessive lateral pressure on the blade. Cygnetic has also been used in maxillo-facial reconstructive surgery, Paediatric pathology and autopsy procedures and for taking nail clippings and biopsies for the diagnosis of infections such as onychomycosis.
Emballering: Supplied individually wrapped in foil packets and boxed in units of 50.