Transfer tubes, Transfertube®
Leverandør: Spectrum Laboratories
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
190195P 190195
SPEC190195PEA 3270 DKK
SPEC190195P SPEC190195
Transfer tubes, Transfertube®
Prøvetagere Væskeprøvetagere
Disposable tubes for liquid and solid samples with one hand. The sample to be transfered is inserted with harvester and discharged using the plunger.
  • Quick and easy use with the finger ring
  • Reduces the risk of cross contamination
Recommended for particulates, suspensions, and microbial colonies.