Liquid sampling dippers
Leverandør: Sampling Systems
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
5005B-500 5005B-250 5005B-100 5005A-100 5005A-250 5005A-500 5005C-100 5005C-250 5005C-500
SAMP5005B-500EA 3000 DKK
SAMP5005B-500 SAMP5005B-250 SAMP5005B-100 SAMP5005A-100 SAMP5005A-250 SAMP5005A-500 300-0461 300-0462 300-0463
Liquid sampling dippers
Prøvetagere Prøveøser
Stainless dippers are ideal for taking surface samples from liquids and suspensions. The sampler has a 316 L stainless steel cup welded to a long reach handle. The handles are made from strong solid stainless steel bar.
  • Range of cup sizes from 100 to 500 ml
  • Lengths from 600 to 1500 mm
  • 316 L stainless steel
  • Polished finish
To take a sample you simply insert the dipper into the liquid to take a surface sample. They are ideal for obtaining samples from a wide range of environments including use in pharmaceutical, chemical and food industries.