Racks and Boxes for Microcentrifuge Tubes
Leverandør: GLW
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
GSTY G50B G50G G50R VST
391-8855EA 38.7 DKK
391-8855 391-8856 391-8857 391-8858 GLWAVST
Racks and Boxes for Microcentrifuge Tubes
Bokse Bokse til Rør
Styro racks are made of heavy expanded polystyrene which are used in freezer down to -90 °C. System box is made of cardboard and is designed to securely enclose two styro rack boxes.
  • Not autoclavable
  • For 1,5 and 2,0 ml tubes up to 12 mm diameter