Stopcock keys, threaded, long for double bore, grounded, DURAN®
Leverandør: DWK Life Sciences
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
286037308 286036706
SCOT286037308EA 225 DKK
SCOT286037308 SCOT286036706
Stopcock keys, threaded, long for double bore, grounded, DURAN®
Stophane nøgler
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance