Capillary columns, Elite-1701 for autosystem
Leverandør: PerkinElmer
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
N9316228 N9316231 N9316248 N9316234 N9316238 N9316243 N9316244 N9316247 N9316255 N9316259
PERCN9316228EA 7370 DKK
PERCN9316228 PERCN9316231 PERCN9316248 PERCN9316234 PERCN9316238 PERCN9316243 PERCN9316244 PERCN9316247 PERCN9316255 PERCN9316259
Capillary columns, Elite-1701 for autosystem
Kromatografikolonner
Elite-1701 (14% Cyanopropylphenyl) Methylpolysiloxane series capillary columns are fused silica capillary columns ideal for the analysis of nonpolar to polar compounds and acid to basic species.