Wipers for general applications, PIG®
Leverandør: NEW PIG
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
WIP235 WIP230 WIP231 WIP310 WIP311
NEWPWIP235EA 770 DKK
NEWPWIP235 NEWPWIP230 NEWPWIP231 NEWPWIP310 NEWPWIP311
Wipers for general applications, PIG®
Papirservietter Engangsservietter og - håndklæder Papirservietter til alment brug
These cellulose wipes with adhesive latex binder are ideal for all kinds of jobs - from cleaning parts to cleaning hands.
  • Constructed of soft, absorbent cellulose fibers with a high-quality binder to increase strength for wet or dry wiping
  • Made thick and absorbent to quickly soak up oils, degreasers and other liquids
  • Low-lint material won't leave residue or create a mess
  • Excellent for a variety of wiping applications from quick cleanups to degreasing
  • Available 1/4 folded and poly wrapped, as economical rolls or in convenient pop-up boxes