VWR®, bufferløsninger, sekundære standarder
Leverandør: VWR Chemicals
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
84582.260EA
429
DKK
84582.260
84583.260
84580.260
84581.260
84584.260
84585.260
84586.260
84587.260
84593.260
VWR®, bufferløsninger, sekundære standarder
Buffere
pH buffere
- Accurate to ±0,010 pH units (except buffer 12,454 with accuracy ±0,050)
- Solution standardised at 25 °C
- Fully traceable for optimum pH calibration (traceable to NIST, DIN 19266 and IUPAC)
- Temperature dependence data is printed on the labels as are batch number and expiry date