Safety canisters with screw cap
Leverandør: DUPERTHAL
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
3.90.005 3.90.010
DUEH3.90.005EA 2570 DKK
DUEH3.90.005 DUEH3.90.010
Safety canisters with screw cap
Sikkerhedsdunke
Safety canister for transport of hazardous substances, made of 1.4571 polished steel with teflon seal.
  • Tight closure, captive cap on flexible shaft
  • Flame arrester in can neck
  • Can be used in-house with measuring tap
  • 38 mm pipe thread
Markings according to GetStoffV and BGV A8, additional markings according to ADR/RID.