Pallets, gas cylinders
Leverandør: DUPERTHAL
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
7.00.800 7.00.810
DUEH7.00.800EA 4540 DKK
DUEH7.00.800 DUEH7.00.810
Pallets, gas cylinders
Vogne
These pallets are used for the safe, economic and effortless transport of 1 to 8 gas cylinders with hot-dip galvanised surface.
  • Sturdy base frame with fortlift pockets
  • Gas cylinders can be secured individually with chains
  • Lateral tilt limiter
  • Hooks for hoses and screw caps
Inner dimensions of fortlift pockets (W×H): 170×70 mm.
Inner distance between pockets: 510 mm.
Certificeringer: According to EN ISO 1461