Retort bases, with M10 thread
Leverandør: NICKEL ELECTRO
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
6890 6891 6892 6893
NICK6890EA 246 DKK
NICK6890 NICK6891 NICK6892 NICK6893
Retort bases, with M10 thread
Retortstativer
Pressed steel finished in matt black textured acrylic paint, threaded M10 × 1,5 mm.
Bestillingsoplysninger: Retort rods are not included, they can be ordered separately under "Complementary products".