Hydrometers A.S.T.M. E100 for mineral oils
Leverandør: Alla France
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
3530LC060/15QP 3530LC065/15QP 3530LC070/15QP 3530LC075/15QP 3530LC080/15QP 3530LC085/15QP 3530LC090/15QP 3530LC095/15QP 3530LC100/15QP 3530LC105/15QP
ALLA3530LC060/15QPEA 681 DKK
ALLA3530LC060/15QP ALLA3530LC065/15QP ALLA3530LC070/15QP ALLA3530LC075/15QP ALLA3530LC080/15QP ALLA3530LC085/15QP ALLA3530LC090/15QP ALLA3530LC095/15QP ALLA3530LC100/15QP ALLA3530LC105/15QP
Hydrometers A.S.T.M. E100 for mineral oils
Flydevægte
The scale of the hydrometer is fully marked with all the reference norms, the calibration temperature, the surface tension, the individual number of the instrument and the serial number.
  • Calibrated at 15 °C, A2LA Traceable hydrometers
  • Graduated in kg/m³, accuracy: 0,5 kg/m³
  • Reading below meniscus