Safety transport containers
Leverandør: ROETZMEIER SICHERH BEHAELTER
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
01T 02T 05T 05KT 10KT 20KTL 10TZ 25TZ 05KTV 10KTV
119-2350EA 1340 DKK
119-2350 119-2351 119-2352 119-2370 119-2371 119-0122 119-0157 119-0156 ROET05KTV 119-0209
Safety transport containers
Sikkerhedsdunke
Safety transport containers for UN approved transport of hazardous goods on public routes.
  • High quality, and chemically resistant stainless steel with PTFE gaskets
  • All types of hazardous goods (UN X approval)
  • Help to avoid non returnable one-way transport packaging
  • Screw cap without pressure control valve - also available with pressure control valve as well
  • Removable flame trap
Design approved according to ADR, RID, IMDG Code, ICAO-TI and GGVSEB.
Certificeringer: Design approved according to ADR, RID, IMDG Code, ICAO-TI and GGVSEB.
Leveringsinformation: 10 and 25 litre barrels, and 20 litre canisters are equipped with separate ventilation screw to maximise the flow rate and allow for decanting in horizontal position, when used in conjunction with self-closing tap (119-2401).