Rotary evaporating flasks, with standard ground joints
Leverandør: witeg Labortechnik
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
0.655.004 0.655.050 0.655.001 0.655.500 0.655.100 0.655.250 0.655.150 0.655.002 0.654.500
531-9128EA 974 DKK
531-9128 WITG0.655.050 WITG0.655.001 WITG0.655.500 WITG0.655.100 WITG0.655.250 WITG0.655.150 WITG0.655.002 WITG0.654.500
Rotary evaporating flasks, with standard ground joints
Kolber Kolber til rotationsfordamper
Borosilicate glass 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Standard ground joint socket, pear-shaped (4000 ml = round bottom), centre neck, thick-walled. Suitable for all standard rotary evaporators; with white labelling area.