Glass stoppers with hexagonal grip
Leverandør: witeg Labortechnik
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
0.302.019 0.303.024 0.303.014 0.303.029 0.302.014
WITG0.302.019EA 31.4 DKK
WITG0.302.019 WITG0.303.024 WITG0.303.014 WITG0.303.029 WITG0.302.014
Glass stoppers with hexagonal grip
Propper Glas
Hollow standard taper stoppers with hexagonal grip, made of borosilicate glass 3.3.
  • Acc. to DIN 12252