Safety devices for conical ground joints, Superfix
Leverandør: witeg Labortechnik
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
0.140.014 0.140.019 0.140.024 0.140.029 0.140.034 0.140.045
201-7211EA 47.2 DKK
201-7211 201-7212 201-7213 201-7214 201-7215 201-7216
Safety devices for conical ground joints, Superfix
Klemmer Klemmer til NS-slib
Safety devices, complete with cap, inner part of thread cone and clamping O-ring. Suitable for cylindrical and conventional connections of conical joints.
  • Solid security
  • Packed in cellophane bags