Cap strips for PCR tubes
Leverandør: EPPENDORF
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
0030124.839 0030124.847
525-0800EA 500 DKK
525-0800 525-0801
Cap strips for PCR tubes
Rør PCR rør
Strips with eight microcaps for 0,1 ml and 0,2 ml wells.
  • Easy and rapid sealing of PCR plates
  • Easy to remove using tab at end of strip
  • With flat or domed shape
Bestil nu
Specifikationer
Environmentally Preferable Lav fremstillingspåvirkning
Miljøcertificering Renewable Energy Certificate, ISO 14001