Magnetic stirring bars, oval
Leverandør: COWIE
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
001.650.17 001.670.27 001.650.RE 001.640.RE 001.635.RE 001.635.13 001.630.10 001.625.RE 001.625.10 001.620.RE 001.615.RE 001.610.RE 001.664.RE 001.640.13 001.6100 001.6150
442-0125EA 76.3 DKK
442-0125 442-0126 COWI001.650.RE COWI001.640.RE COWI001.635.RE 442-0124 442-0123 COWI001.625.RE 442-0122 COWI001.620.RE COWI001.615.RE COWI001.610.RE COWI001.664.RE COWI001.640.13 COWI001.6100 COWI001.6150
Magnetic stirring bars, oval
Omrørere Omrøremagneter
PTFE covered. For stirrers with rare earth magnetic core the magnetic attraction is 1,5 to 2 times stronger than conventional stirrers. The magnetic core is made from a combination of samarium and cobalt. The rare earth stirrers will continue to function at full strength for many years as the magnetism remains for longer.
  • Very good chemical resistance
  • Temperature resistant, suitable for use at high and low temperatures (−200 to +280 °C)