Tubing, PTFE
Leverandør: COWIE
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
016.1706 016.1708 016.1714 016.1712
COWI016.1706EA 172 DKK
COWI016.1706 COWI016.1708 COWI016.1714 COWI016.1712
Tubing, PTFE
Slanger Overførselsslange
PTFE, white.
  • Excellent chemical resistance
  • Very good thermal resistance, autoclavable
  • Free of extractable substances, physiologically neutral, biocompatible
This tubing is characterised by its non stick surface and excellent dielectric properties.
Temperature resistance: −200 to +260 °C