Tubing connectors, straight through, Bayonet type
Leverandør: COWIE
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
016.905.2 016.904.9.2 016.904.2 016.903.7.2 016.903.2 016.902.5.2 016.902.2
COWI016.905.2EA 169 DKK
COWI016.905.2 COWI016.904.9.2 COWI016.904.2 COWI016.903.7.2 COWI016.903.2 COWI016.902.5.2 COWI016.902.2
Tubing connectors, straight through, Bayonet type
Fittings og stik
Straight through connectors, made of PTFE, bayonet type for flexible tubings.
  • Very good chemical resistance
  • Very good temperature resistance