Volumetric flasks, class A, white markings, USP-individual certificate, BLAUBRAND®
Leverandør: Brand
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
USP37464 USP37467 USP37465 USP37463 USP37462 USP37461 USP37469 USP37490 USP37489 USP37488 USP37487 USP37485 USP37484 USP37483 USP37482 USP37481 USP37470 USP37491 USP37468 USP37471
BRNDUSP37464EA 714 DKK
BRNDUSP37464 BRNDUSP37467 BRNDUSP37465 BRNDUSP37463 BRNDUSP37462 BRNDUSP37461 BRNDUSP37469 BRNDUSP37490 BRNDUSP37489 BRNDUSP37488 BRNDUSP37487 BRNDUSP37485 BRNDUSP37484 BRNDUSP37483 BRNDUSP37482 BRNDUSP37481 BRNDUSP37470 BRNDUSP37491 BRNDUSP37468 BRNDUSP37471
Volumetric flasks, class A, white markings, USP-individual certificate, BLAUBRAND®
Kolber Volumetriske kolber
Borosilicate glass 3.3, amber, with PP or glass stopper.
  • Calibrated to contain (TC, IN)
  • Mark and inscriptions in high contrast white enamel
  • DE-M marking
  • Supplied with USP-individual certificate
*Wide neck
Certificeringer: DIN EN ISO 1042