AEBSF hydrochloride (4-(2-aminoethyl)benzenesulphonylfluoride hydrochloride), Supelco®
Leverandør: Merck
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Advarsel
Synonymer: 4-(2-Aminoethyl)benzenesulphonyl fluoride hydrochloride, AEBSF hydrochloride
M-Clarity™ quality level = MQ200
76307-100MG 76307-500MG 76307-1G
76307-100MGEA 961 DKK
76307-100MG 76307-500MG 76307-1G
AEBSF hydrochloride (4-(2-aminoethyl)benzenesulphonylfluoride hydrochloride), Supelco®
AEBSF hydrochloride (4-(2-aminoethyl)benzenesulphonylfluoride hydrochloride)
Formel: C₈H₁₁ClFNO₂S
Molekylvægt: 239.69 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00132962
CAS-nummer: 30827-99-7
UN nummer: 3261
ADR: 8 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=846460b7a7e9c74a0f0745dd3b3362950490e0ae&vid=9550947342b3fd3e935d9ef8f10b1d6a48956bb7&prid=web&clid=SAPDAM