Safety harness, Revolution Comfort R2
Leverandør: HONEYWELL SAFETY
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1014237 1014236
113-1272EA 3110 DKK
113-1272 139-0239
Safety harness, Revolution Comfort R2
Faldbeskyttelse Faldseler
Two-point safety harness for use with fall protection equipment and a suitable anchorage point. The webbing is made from DuraFlex®, a highly elastic stretch fabric. Thanks to its unique technology and versatility, the harness is ideally suited for scaffolding, steel and mast construction applications, as well as for general use and maintenance.
  • Safety loop at the back (D-ring) and the front (webbing loops)
  • PivotLink connection at waist height for increased mobility and comfort thanks to optimum ergonomics when bending and stretching
  • Single-handed adjustment thanks to cam buckles and harness clips; automatic quick-lock mechanisms
  • ErgoArmor™ padded back shield for added comfort
  • Integrated accessory system: Pockets for tools and accessories can easily be fastened to or removed from the modular attachment system
EN 361
Certificeringer: EN 361