Safety helmets, 1100
Leverandør: CENTURION SAFETY PRODUCTS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
S01EYC S01EBRC S01EYRC S01BA S01WA S01YA S01CBC
111-0854EA 0 DKK
111-0854 111-1675 CENUS01EYRC 111-1502 111-1512 111-1513 111-1616
Safety helmets, 1100
Hjelme Sikkerhedshjelme
Protective helmet with helmet shell made from high density polyethylene (HDPE). For use in construction and industry.
  • With six point LDPE inner cradle
  • Ear protection, face protection shield and chin strap available as accessories
Size: 510 to 630 mm
Weight: 375 g
EN 397, 440 V, –10 °C, lateral deformation (LD), molten metal (MM)
Certificeringer: EN 397, 440 V, –10 °C, lateral deformation (LD), molten metal (MM)