VOA vials with septa caps
Leverandør: EP SCIENTIFIC
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
S236-0040 S146-0040 S226-0020 S326-0020 S336-0040 S126-0020 S136-0040
EPSCS236-0040EA 2160 DKK
EPSCS236-0040 EPSCS146-0040 EPSCS226-0020 EPSCS326-0020 EPSCS336-0040 EPSCS126-0020 EPSCS136-0040
VOA vials with septa caps
Hætteglas
These vials made of type I borosilicate glass are designed for water sample collection for volatile organic analysis (VOA). Available with septum permanently bonded to the cap or unbonded for easy replacement.
  • Perfect shoulder angle enables effortless filling without air entrapment
  • Open-top white PP cap with standard 3,2 mm thick PTFE lined silicone septum
  • Septum unbonded for easy replacement
  • Directly compatible with automation cosures assembled to vials
Certified vials meet exceed EPA performance-based standards for volatile organic analysis. Certificate of Analysis (CofA) included in the case and bardcoded with lot number for traceability. Each vial individually barcoded with lot number and unique container number. Carton sealed with security tape providing tamper evidence. Chipboard divider packaging to cushion and help protect vials from breakage.
Processed vials are processed in the same manner as certified vials and come with all the features of certified vials, but vials are not barcoded and CofA is not included.
Unprocessed vials are economically packaged glass-to-glass in a shrink-wrapped tray and are ready for own in-house cleaning and certification process.