Chlortetracyclin, HCl

Leverandør: Serva
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Chlortetracycline HCI, Aureomycin, 7-Chlorotetracycline hydrochloride, (4S,4aS,5aS,6S,12aS)-7-Chloro-4-(dimethylamino)-3,6,10,12,12a-pentahydroxy-6-methyl-1,11-dioxo-1,4,4a,5,5a,6,11,12a-octahydrotetracene-2-carboxamide hydrochloride

14200.01 14200.02
SERA14200.01EA 0 DKK
SERA14200.01 SERA14200.02
Chlortetracyclin, HCl
Chlortetracyclin, HCl
Formel: C₂₂H₂₃ClN₂O₈·HCl
Opbevaringstemperatur: Køleskab
MDL Number: MFCD26142699
CAS-nummer: 64-72-2
EINECS: 200-591-7

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR