Ampicillin, natriumsalt

Leverandør: Serva
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: D(-)-α-Aminobenzylpenicillin Sodium Salt, Ampicillin Na, Ampicillin sodium salt, D-(−)-α-Aminobenzylpenicillin sodium salt Ampicillin sodium salt

13399.01 13399.02 13399.03
SERA13399.01EA 320.96 DKK
SERA13399.01 SERA13399.02 SERA13399.03
Ampicillin, natriumsalt
Ampicillin, natriumsalt
Formel: C₁₆H₁₈N₃NaO₄S
Molekylvægt: 371.39 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00064313
CAS-nummer: 69-52-3
EINECS: 200-708-1

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR