Dextran, M.W. ~ 100,000

Leverandør: Serva
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

18693.01 18693.02
SERA18693.01EA 0 DKK
SERA18693.01 SERA18693.02
Dextran, M.W. ~ 100,000
Dextran
Formel: (C₆H₁₀O₅)n
MDL Number: MFCD00130935
CAS-nummer: 9004-54-0
EINECS: 232-677-5

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR