Ball valves for barrels, LaboPlast®
Leverandør: BURKLE
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
0500-8003 0500-8001 0500-0134 0500-8002
227-1108EA 1100 DKK
227-1108 227-1109 BURK0500-0134 BURK0500-8002
Ball valves for barrels, LaboPlast®
Stophaner
PP, black/red
  • Suitable for solidifying media
  • Max. 4 bar pressure at 20 °C
  • Individually adaptable, through various accessories
Ball valves for barrels with thread connections.