Tubing connectors, SP Bel-Art
Leverandør: Bel-Art Products, a Part of SP
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
H197270000 H197280000 H197290000
129-2235EA 1260 DKK
129-2235 129-2236 129-2237
Tubing connectors, SP Bel-Art
Fittings og stik
Unit consists of two barbed PE connectors, which assemble tightly together with a male-female centre taper.
  • Suitable for vacuum applications
  • Temperature resistant up to 80 °C
  • Good chemical reistance