Lyophilisation ampoules, pre-scored, Vacule®, WHEATON®
Leverandør: DWK Life Sciences
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
651502 W651506
WHEA651502EA 2320 DKK
WHEA651502 WHEAW651506
Lyophilisation ampoules, pre-scored, Vacule®, WHEATON®
Ampuller
Made from low extractable borosilicate glass
  • Pre-scored for easy opening
  • Heat sealed only
These ampoules are ideal for small volume lyophilisation samples.
Certificeringer: USP Type I, ASTM E 438 Type I, Class A