Ampoules, pre-scored, WHEATON®
Leverandør: DWK Life Sciences
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
176779 176792 176796 176799 176772 176776 176780 176782
215-8008EA 1490 DKK
215-8008 215-4536 215-4537 215-4538 215-4532 215-4533 215-4534 215-4535
Ampoules, pre-scored, WHEATON®
Ampuller
Glass ampoules made from low extractable borosilicate glass can be hermetically sealed to preserve purity and extend content shelf life.
  • Tamper-evident seal
  • For biological product preservation
  • Gold band on the stem of the ampoule indicates that it is pre-scored to facilitate snapping off the top eliminating the need to file the ampoule
Ideal for standards packaging, environmental and diagnostic standards, lyophilisation and cryogenic storage.
Certificeringer: USP Type I, ASTM E438 Type I, Class A