Test tube racks, Origami TubeRack
Leverandør: TENAK
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
TE50510 TE50530 TE50550
211-0387EA 292 DKK
211-0387 211-0388 211-0389
Test tube racks, Origami TubeRack
Stativer Tube Racks
PP
  • Height adjustable which makes it very flexible for a wide range of tubes
  • Alphanumeric grid for easy location of the samples
  • Can be used to store samples from –45 to 60 °C
An innovative design that is delivered flat packed to reduce shipping cost and storage space. Easy to assemble.