VWR®, Protease inhibitor cocktails, general use
Leverandør: VWR Chemicals
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
M221-1MLEA 750 DKK
M221-1ML M222-1ML
VWR®, Protease inhibitor cocktails, general use
Celledyrkningssupplementer og additiver
Contains a mixture of protease inhibitors: AEBSF, Aprotinin, E-64, Bestatin and Leupeptin. Each vial can be reconstituted in 1 ml deionised water to form a 100X solution.
  • Effectively preserves the native state of proteins
M222 with EDTA