S-Ethylisothiouroniumbromid

Leverandør: Cayman Chemical
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Ethyl carbamimidothioate hydrobromide, S-Ethylisothiourea Hydrobromide, S-EIT hydrobromide, 2-Ethyl-2-thiopseudourea hydrobromide

81275-100 81275-50 81275-500 81275-1
CAYM81275-100EA 631 DKK
CAYM81275-100 CAYM81275-50 CAYM81275-500 CAYM81275-1
S-Ethylisothiouroniumbromid
S-Ethylisothiouronium bromide
Formel: C₃H₉BrN₂S
MDL Number: MFCD00012585
CAS-nummer: 1071-37-0

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR