γ-Cyclodextrin, hydrat 99%
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer: γ-Cyclodextrin hydrate, γ-Cyclodextrin xhydrate
229900010 229905000 229900050
ACRO229900010EA 1900 DKK
ACRO229900010 ACRO229905000 ACRO229900050
γ-Cyclodextrin, hydrat 99%
γ-Cyclodextrin, hydrat
Formel: C₄₈H₈₀O₄₀·H₂O
Molekylvægt: 1297.15 g/mol (anhy)
Smeltepunkt: 267 °C (dec.)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00149574
CAS-nummer: 91464-90-3
EINECS: 241-482-4
Bestil nu
Specifikationer Testresultater
Appearance White to off-white Powder
Infrared spectrum Conforms
Water ≤11 %
HPLC ≥98.5 % (ELSD)