6-Bromisoquinolin

Leverandør: Cayman Chemical
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

10007080-1 10007080-500
CAYM10007080-1EA 1220 DKK
CAYM10007080-1 CAYM10007080-500
6-Bromisoquinolin
6-Bromisoquinolin
Formel: C₉H₆BrN
MDL Number: MFCD04973299
CAS-nummer: 34784-05-9

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR