Methallyltributyltin ≥98%
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Farlig
L20186.03 L20186.06
L20186.03EA 965 DKK
L20186.03 L20186.06
Methallyltributyltin ≥98%
Methallyltributyltin
Purity: 98% CAS No.: 67883-62-9 Molecular Formula: C16H34Sn Formula Weight: 345.16 Refractive Index: 1.4875
Formel: C₁₆H₃₄Sn
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD03425867
CAS-nummer: 67883-62-9
UN nummer: 2788
ADR: 6.1,II