(R)-(-)-Methyl3-hydroxybutyrat ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: (R)-Methyl 3-hydroxybutanoate, Methyl (R)-3-hydroxybutyrate, Methyl (R)-(-)-3-Hydroxybutyrate

H0705-10ML H0705-25ML
TCIAH0705-10MLEA 140 DKK
TCIAH0705-10ML TCIAH0705-25ML
(R)-(-)-Methyl3-hydroxybutyrat ≥98.0% (by GC)
(R)-(-)-Methyl3-hydroxybutyrat
Formel: C₅H₁₀O₃
Kogepunkt: 75°C
Flammepunkt: 73°C
MDL Number: MFCD00063289
CAS-nummer: 3976-69-0
EINECS: 223-610-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=13dc94dbf83df60056cfc77889e95d4aa128702d&vid=13dc94dbf83df60056cfc77889e95d4aa128702d&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Specific rotation [a]20/D -21.0 to -24.0 deg (neat)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR