Robot-spidser til Agilent
Leverandør: BIOTIX
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
VB-0030-3SC 63300094 63300093 63300092
736-0409EA 6580 DKK
736-0409 736-0413 736-0614 BITX63300092
Robot-spidser til Agilent
Pipettespidser
Biotix robotic tips for Agilent™ VPrep® and Bravo® platforms are engineered to ensure complete compatibility, requiring no changes in programming scripts in the platform's liquid handling software suite.
  • Made of medical-grade virgin PP
  • Tips are BioReady™ certified to be free of RNase, DNase, and Endotoxins (pyrogens)
  • Available as standard or filtered tips
Manufactured to the highest engineering standards for robotic systems, the ultra-straight tips target the centre of microplate wells precisely, delivering exceptional accuracy and precision without risk of jamming.