Capillary columns, Velocity-1
Leverandør: PerkinElmer
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
N9306318 N9306321
568-0281EA 13070 DKK
568-0281 568-0282
Capillary columns, Velocity-1
Kromatografikolonner
General purpose columns with a highly versatile phase that is extremely rugged, exhibiting long column lifetime and high operating temperatures. Ideal for the analysis of non polar petrochemical samples, such as detailed hydrocarbon analysis, hydrocarbon gases, petroleum oxygenates, petroleum aromatics, fuels waxes, oils, sulphur compounds, mercaptains and carbon disulphide. It also an excellent phase for solvents, chemicals, flavours, fragrances, essential oils, air toxins, chlorofluorocarbons, pesticides and hydrocarbons.
100% dimethyl polysiloxane
Max. temperature: 350 °C