Cellulose stoppers, Steristoppers®
Leverandør: HERENZ HEINZ
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1012350 1011000 1011201 1011800 1013800 1014001 1013600 1011300 1011450 1011500 1011801 1010700 1011200 1011250 1011406 1012150 1011900 1011410 1012721 1011400 1010906 1011501 1012905 1010715 1012919 1011100 1013225 1013201 1011409 1011806 1010502 1012205 1013700 1012723 1010800
217-9935EA 14430 DKK
217-9935 391-8908 391-8909 391-8922 391-8950 217-9951 391-8949 391-8910 217-9919 391-8920 HERE1011801 217-9907 217-9909 217-9911 217-9916 217-9936 217-9937 217-9941 391-8970 HERE1011400 HERE1010906 HERE1011501 HERE1012905 HERE1010715 HERE1012919 HERE1011100 HERE1013225 HERE1013201 HERE1011409 HERE1011806 HERE1010502 HERE1012205 HERE1013700 HERE1012723 HERE1010800
Cellulose stoppers, Steristoppers®
Propper Cellulosepropper
Original Steristoppers® have been proved as an excellent sealing for microbiological samples and tissue cultures in test tubes and Erlenmeyer flasks. They are in constant use in many world famous microbiological laboratories.
  • Bacteria filtering
  • Air permeable
  • For single use
  • Available from 4 to 60 mm Ø