VWR®, Tunics with Collar, Polyester/Cotton
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
113-1855EA 411 DKK
113-1855 113-1854 113-1853 113-1852 113-1851 113-1850 113-1849 113-1848 113-1847 113-1846 113-1870 113-1866 113-1867 113-1869 113-1871 113-1872 113-1873 113-1874 113-1875 113-1868 113-5995 113-6180
VWR®, Tunics with Collar, Polyester/Cotton
Arbejdstøj
These functional tunics with a tailored collar are made of 65% polyester and 35% cotton. They are suitable for use in medical areas, especially in medical care.
  • Two side pockets (one with an additional small pocket) and one breast pocket
  • Concealed press stud fastening with five snap buttons
  • With side slits for improved wearing comfort
  • Washable at 95 °C, suitable for industrial laundering
Colour: White
Density: 215 g/m²
Approved in accordance with Oeko-Tex® Standard 100.
Certificeringer: Approved in accordance with Oeko-Tex® Standard 100.