Safety shields, SP Bel-Art
Leverandør: Bel-Art Products, a Part of SP
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
H249640000 H249620000
111-2604EA 12780 DKK
111-2604 111-2603
Safety shields, SP Bel-Art
Afskærmning Sikkerhedsafskærmninger
The stable transparent safety shield PC (thickness: 4,7 mm) is constructed of clear PC barrier and heavy steel base is coated with blue epoxy coating.
  • Three-sided construction provides front and side protection against flying debris
  • Plated steel bolts, nuts and washers ensure long life and rust-resistance
Certificeringer: ANSI Z87.1, 9.11, 9.12